4-Acetyl-1H-pyrrole-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

042740-500MG 042740-1G 042740-5G
101857-272EA 132.31 USD
101857-272 101857-274 101857-276
4-Acetyl-1H-pyrrole-2-carboxylic acid ≥97%
4-Acetyl-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₇NO₃
Melting Pt: 229…232 °C
MDL Number: MFCD02571849
CAS Number: 16168-93-7

Order Now

Specification Test Results

Melting point 229-232°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR