3-[5-Chloro-2-(2-propynyloxy)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

042739-500MG 042739-1G 042739-5G
101857-266EA 375.38 USD
101857-266 101857-268 101857-270
3-[5-Chloro-2-(2-propynyloxy)phenyl]acrylic acid ≥95%
3-[5-Chloro-2-(2-propynyloxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₂H₉ClO₃
Melting Pt: 155…158 °C
MDL Number: MFCD03001216
CAS Number: 695186-82-4

Order Now

Specification Test Results

Melting point 155-158°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR