2,5-Difluoro-4-nitrobenzoic acid ≥95%
Supplier: Matrix Scientific
042738-1G
042738-5G
101857-262EA
20
USD
101857-262
101857-264
2,5-Difluoro-4-nitrobenzoic acid ≥95%
2,5-Difluoro-4-nitrobenzoic acid
Hazard: Irritant
Formula:
C₇H₃F₂NO₄ MW: 203.10 g/mol Melting Pt: 147-148°C |
MDL Number:
MFCD03425705 CAS Number: 116465-48-6 |
Specification Test Results
Melting point | 147-148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...