tert-Butyl-2-oxo-1-oxa-8-azaspiro[4.5]decane-8-carboxylate ≥95%

Supplier: Matrix Scientific

042733-500MG 042733-1G
101857-246EA 120 USD
101857-246 101857-248
tert-Butyl-2-oxo-1-oxa-8-azaspiro[4.5]decane-8-carboxylate ≥95%
tert-Butyl-2-oxo-1-oxa-8-azaspiro[4.5]decane-8-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₁NO₄
MW: 255.32 g/mol
Melting Pt: 125…127 °C
MDL Number: MFCD11100196
CAS Number: 301226-27-7

Order Now

Specification Test Results

Melting point 125-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR