6,8-Dibromo-2-oxo-2H-chromene-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

042732-500MG 042732-1G 042732-5G
101857-240EA 181.54 USD
101857-240 101857-242 101857-244
6,8-Dibromo-2-oxo-2H-chromene-3-carboxylic acid ≥95%
6,8-Dibromo-2-oxo-2H-chromene-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₄Br₂O₄
Melting Pt: 218…220 °C
MDL Number: MFCD00173735
CAS Number: 3855-87-6

Order Now

Specification Test Results

Melting point 218-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR