6-Methyl-1H-indazol-5-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Amino-6-methyl-1H-indazole
042731-500MG
042731-1G
042731-5G
101857-234EA
206.15
USD
101857-234
101857-236
101857-238
6-Methyl-1H-indazol-5-amine ≥95%
6-Methyl-1H-indazol-5-amine
Hazard: Irritant/Refrigerate
Formula:
C₈H₉N₃ MW: 147.18 g/mol Melting Pt: 215…217 °C |
MDL Number:
MFCD03305372 CAS Number: 81115-45-9 |
Specification Test Results
Melting point | 215-217°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...