6-Methyl-5-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

042730-500MG 042730-1G 042730-5G
101857-228EA 69.23 USD
101857-228 101857-230 101857-232
6-Methyl-5-nitro-1H-indazole ≥95%
6-Methyl-5-nitro-1H-indazole

Hazard: Irritant

Formula: C₈H₇N₃O₂
MW: 177.1601 g/mol
Melting Pt: 227-230°C
MDL Number: MFCD03305368
CAS Number: 81115-43-7

Order Now

Specification Test Results

Melting point 227-230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR