1-Phenyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

042725-500MG 042725-1G 042725-5G
101857-206EA 278.46 USD
101857-206 101857-208 101857-210
1-Phenyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylic acid ≥95%
1-Phenyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₁N₃O₂
Melting Pt: 185…189 °C
MDL Number: MFCD01749393
CAS Number: 116834-08-3

Order Now

Specification Test Results

Melting point 185-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR