Ethyl-1-phenyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

042724-500MG 042724-1G 042724-5G
101857-200EA 278.46 USD
101857-200 101857-202 101857-204
Ethyl-1-phenyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-phenyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₅N₃O₂
Melting Pt: 113…115 °C
MDL Number: MFCD02571083
CAS Number: 94692-05-4

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR