4-(Phenoxymethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

042723-500MG 042723-1G 042723-5G
101857-194EA 347.69 USD
101857-194 101857-196 101857-198
4-(Phenoxymethyl)benzoic acid ≥95%
4-(Phenoxymethyl)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₃
MW: 228.2433 g/mol
Melting Pt: 223…226 °C
MDL Number: MFCD00623268
CAS Number: 31719-76-3

Order Now

Specification Test Results

Melting point 223-226°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR