Ethyl-3-[4-(tert-butyl)phenyl]-2-cyanoacrylate ≥95%

Supplier: Matrix Scientific

042715-500MG 042715-1G
101857-154EA 181.54 USD
101857-154 101857-156
Ethyl-3-[4-(tert-butyl)phenyl]-2-cyanoacrylate ≥95%
Ethyl-3-[4-(tert-butyl)phenyl]-2-cyanoacrylate

Hazard: Irritant

Formula: C₁₆H₁₉NO₂
MW: 257.34 g/mol
Melting Pt: 67…69 °C
MDL Number: MFCD04241676
CAS Number: 247099-46-3

Order Now

Specification Test Results

Melting point 67-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR