N'-(4-Formyl-3-methyl-1-phenyl-1H-pyrazol-5-yl)-N,N-dimethyliminoformamide ≥95%

Supplier: Matrix Scientific

042712-500MG 042712-1G 042712-5G
101857-136EA 375.38 USD
101857-136 101857-138 101857-140
N'-(4-Formyl-3-methyl-1-phenyl-1H-pyrazol-5-yl)-N,N-dimethyliminoformamide ≥95%
N'-(4-Formyl-3-methyl-1-phenyl-1H-pyrazol-5-yl)-N,N-dimethyliminoformamide

Hazard: Irritant

Formula: C₁₄H₁₆N₄O
MW: 256.31 g/mol
Melting Pt: 86…89 °C
MDL Number: MFCD03152811
CAS Number: 58668-41-0

Order Now

Specification Test Results

Melting point 86-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR