2-Chlorothiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-formyl-1,3-thiazole, 2-chloro-1,3-thiazole-5-carbaldehyde, 2-Chlorothiazole-5-carboxaldehyde

042709-1G 042709-5G
101857-132EA 38.46 USD
101857-132 101857-134
2-Chlorothiazole-5-carbaldehyde ≥95%
2-Chlorothiazole-5-carbaldehyde

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₄H₂ClNOS
MW: 147.58 g/mol
Melting Pt: 85…88 °C
MDL Number: MFCD01568717
CAS Number: 95453-58-0

Order Now

Specification Test Results

Melting point 85-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR