4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-N'-hydroxybenzenecarboximidamide ≥95%

Supplier: Matrix Scientific

042708-500MG 042708-1G 042708-5G
101857-124EA 375.38 USD
101857-124 101857-126 101857-128
4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-N'-hydroxybenzenecarboximidamide ≥95%
4-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃O₂S
Melting Pt: 179…181 °C
MDL Number: MFCD01568715
CAS Number: 341967-76-8

Order Now

Specification Test Results

Melting point 179-181°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR