4-(4-Methylpiperazino)-3-nitrobenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

042706-500MG 042706-1G 042706-5G
101857-112EA 556.92 USD
101857-112 101857-114 101857-116
4-(4-Methylpiperazino)-3-nitrobenzenecarboxylic acid ≥95%
4-(4-Methylpiperazino)-3-nitrobenzenecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₄
Melting Pt: 264 °C
MDL Number: MFCD01568172
CAS Number: 137726-00-2

Order Now

Specification Test Results

Melting point 264°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR