Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarboxylate ≥97%
Supplier: Matrix Scientific
042705-500MG
042705-1G
042705-5G
101857-106EA
278.46
USD
101857-106
101857-108
101857-110
Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarboxylate ≥97%
Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarboxylate
Hazard: Irritant
Formula:
C₁₄H₉ClF₃NO₃ Melting Pt: 79…81 °C |
MDL Number:
MFCD01568618 CAS Number: 341967-62-2 |
Specification Test Results
Melting point | 79-81°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...