2-(4-Methylphenyl)-1-hydrazinecarbothioamide ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzenecarboxylate

042704-500MG 042704-1G 042704-5G
101857-100EA 278.46 USD
101857-100 101857-102 101857-104
2-(4-Methylphenyl)-1-hydrazinecarbothioamide ≥95%
2-(4-Methylphenyl)-1-hydrazinecarbothioamide

Hazard: Irritant

Formula: C₈H₁₁N₃S
MW: 181.26 g/mol
Melting Pt: 172…173 °C
MDL Number: MFCD03791137
CAS Number: 7382-40-3

Order Now

Specification Test Results

Melting point 172-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR