3-Hydroxy-6-methyl-2,3-dihydro-4H-furo[3,2-c]pyran-4-one ≥95%

Supplier: Matrix Scientific

042703-500MG 042703-1G 042703-5G
101857-094EA 278.46 USD
101857-094 101857-096 101857-098
3-Hydroxy-6-methyl-2,3-dihydro-4H-furo[3,2-c]pyran-4-one ≥95%
3-Hydroxy-6-methyl-2,3-dihydro-4H-furo[3,2-c]pyran-4-one

Hazard: Irritant

Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 137…139 °C
MDL Number: MFCD01443822
CAS Number: 57053-18-6

Order Now

Specification Test Results

Melting point 137-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR