Ethyl-6-oxo-2-phenyl-1,6-dihydro-5-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 1,6-dihydro-6-oxo-2-phenylpyrimidine-5-carboxylate
042701-500MG
042701-1G
042701-5G
101857-082EA
58.46
USD
101857-082
101857-084
101857-086
Ethyl-6-oxo-2-phenyl-1,6-dihydro-5-pyrimidinecarboxylate ≥95%
Ethyl-6-oxo-2-phenyl-1,6-dihydro-5-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₃ MW: 244.25 g/mol Melting Pt: 209…212 °C |
MDL Number:
MFCD00695462 CAS Number: 55613-22-4 |
Specification Test Results
Melting point | 209-212°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...