Ethyl-3-(4-chlorophenyl)-5-hydroxy-1,2,4-triazine-6-carboxylate ≥95%

Supplier: Matrix Scientific

042700-500MG 042700-1G 042700-5G
101857-076EA 747.69 USD
101857-076 101857-078 101857-080
Ethyl-3-(4-chlorophenyl)-5-hydroxy-1,2,4-triazine-6-carboxylate ≥95%
Ethyl-3-(4-chlorophenyl)-5-hydroxy-1,2,4-triazine-6-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O₃
Melting Pt: 247…250 °C
MDL Number: MFCD04117779
CAS Number: 36286-77-8

Order Now

Specification Test Results

Melting point 247-250°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR