1-[4-(Methylsulfanyl)phenyl]-1,4-pentanedione ≥95%

Supplier: Matrix Scientific

042697-500MG 042697-1G
101857-066EA 278.46 USD
101857-066 101857-068
1-[4-(Methylsulfanyl)phenyl]-1,4-pentanedione ≥95%
1-[4-(Methylsulfanyl)phenyl]-1,4-pentanedione

Hazard: Irritant

Formula: C₁₂H₁₄O₂S
MW: 222.31 g/mol
Melting Pt: 75…78 °C
MDL Number: MFCD08056601
CAS Number: 189501-33-5

Order Now

Specification Test Results

Melting point 75-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR