3-Chloro-N-[4-(dimethylamino)phenyl]-2,2-dimethylpropanamide ≥95%
Supplier: Matrix Scientific
042694-500MG
042694-1G
042694-5G
101857-048EA
181.54
USD
101857-048
101857-050
101857-052
3-Chloro-N-[4-(dimethylamino)phenyl]-2,2-dimethylpropanamide ≥95%
3-Chloro-N-[4-(dimethylamino)phenyl]-2,2-dimethylpropanamide
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O Melting Pt: 104…106 °C |
MDL Number:
MFCD00793368 CAS Number: 341966-00-5 |
Specification Test Results
Melting point | 104-106°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...