5-[(4-Methoxyphenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

042693-500MG 042693-1G 042693-5G
101857-042EA 747.69 USD
101857-042 101857-044 101857-046
5-[(4-Methoxyphenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde ≥95%
5-[(4-Methoxyphenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₈H₁₆N₂O₂S
Melting Pt: 94…98 °C
MDL Number: MFCD00975682
CAS Number: 321998-19-0

Order Now

Specification Test Results

Melting point 94-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR