[3-(4-Chlorophenyl)-5-isoxazolyl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
(3-(4-Chlorophenyl)isoxazol-5-yl)methanol
042692-500MG
042692-1G
042692-5G
101857-036EA
320
USD
101857-036
101857-038
101857-040
[3-(4-Chlorophenyl)-5-isoxazolyl]methanol ≥95%
[3-(4-Chlorophenyl)-5-isoxazolyl]methanol
Hazard: Irritant
Formula:
C₁₀H₈ClNO₂ MW: 209.6290 g/mol Melting Pt: 98…100 °C |
MDL Number:
MFCD01935957 CAS Number: 206055-90-5 |
Specification Test Results
Melting point | 98-100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...