5-Amino-1-(tert-butyl)-3-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

042689-500MG 042689-1G 042689-5G
101857-018EA 190.77 USD
101857-018 101857-020 101857-022
5-Amino-1-(tert-butyl)-3-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-(tert-butyl)-3-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₄H₁₅ClN₄
Melting Pt: 154…156 °C
MDL Number: MFCD01935935
CAS Number: 180903-14-4

Order Now

Specification Test Results

Melting point 154-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR