3-Amino-4-[(4-methylphenyl)sulfanyl]benzenecarbonitrile ≥95%
Supplier: Matrix Scientific
042684-500MG
042684-1G
042684-5G
101856-992EA
278.46
USD
101856-992
101856-994
101856-996
3-Amino-4-[(4-methylphenyl)sulfanyl]benzenecarbonitrile ≥95%
3-Amino-4-[(4-methylphenyl)sulfanyl]benzenecarbonitrile
Hazard: Irritant
Formula:
C₁₄H₁₂N₂S MW: 240.33 g/mol Melting Pt: 98…101 °C |
MDL Number:
MFCD00793310 CAS Number: 337924-03-5 |
Specification Test Results
Melting point | 98-101°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...