3-Amino-4-(phenylsulfanyl)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

042683-500MG 042683-1G 042683-5G
101856-986EA 375.38 USD
101856-986 101856-988 101856-990
3-Amino-4-(phenylsulfanyl)benzenecarbonitrile ≥95%
3-Amino-4-(phenylsulfanyl)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₁₀N₂S
Melting Pt: 103…106 °C
MDL Number: MFCD00793304
CAS Number: 337923-85-0

Order Now

Specification Test Results

Melting point 103-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR