{2-[(4-Chlorophenyl)sulfanyl]phenyl}methanol ≥95%

Supplier: Matrix Scientific

042681-500MG 042681-1G 042681-5G
101856-974EA 278.46 USD
101856-974 101856-976 101856-978
{2-[(4-Chlorophenyl)sulfanyl]phenyl}methanol ≥95%

Hazard: Irritant

Formula: C₁₃H₁₁ClOS
Melting Pt: 34…37 °C
MDL Number: MFCD04108002
CAS Number: 13459-59-1

Order Now

Specification Test Results

Melting point 34-37°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR