Methyl-5-cyano-6-hydroxy-2-methylnicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-cyano-6-hydroxy-2-methylnicotinate
042679-500MG
042679-1G
042679-5G
101856-964EA
224.62
USD
101856-964
101856-966
101856-968
Methyl-5-cyano-6-hydroxy-2-methylnicotinate ≥95%
Methyl-5-cyano-6-hydroxy-2-methylnicotinate
Hazard: Irritant
Formula:
C₉H₈N₂O₃ Melting Pt: >250 °C (dec.) |
MDL Number:
MFCD00793171 CAS Number: 71408-02-1 |
Specification Test Results
Melting point | >250°C(dec) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...