5-Acetyl-2-chloro-6-methylnicotinonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Acetyl-2-chloro-6-methylpyridine-3-carbonitrile

042678-500MG 042678-1G 042678-5G
101856-958EA 556.92 USD
101856-958 101856-960 101856-962
5-Acetyl-2-chloro-6-methylnicotinonitrile ≥97%
5-Acetyl-2-chloro-6-methylnicotinonitrile

Hazard: Irritant

Formula: C₉H₇ClN₂O
MW: 194.62 g/mol
Melting Pt: 81…83 °C
MDL Number: MFCD01315263
CAS Number: 121348-15-0

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR