5-Acetyl-2-hydroxy-6-methylnicotinonitrile ≥95%

Supplier: Matrix Scientific

042676-500MG 042676-1G 042676-5G
101856-946EA 135.38 USD
101856-946 101856-948 101856-950
5-Acetyl-2-hydroxy-6-methylnicotinonitrile ≥95%
5-Acetyl-2-hydroxy-6-methylnicotinonitrile

Hazard: Irritant

Formula: C₉H₈N₂O₂
Melting Pt: 233…235 °C
MDL Number: MFCD00084860
CAS Number: 52600-53-0

Order Now

Specification Test Results

Melting point 233-235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR