1-(Dimethylamino)-5-(3-methoxyphenyl)-1,4-pentadien-3-one ≥95%
Supplier: Matrix Scientific
042675-500MG
042675-1G
042675-5G
101856-940EA
181.54
USD
101856-940
101856-942
101856-944
1-(Dimethylamino)-5-(3-methoxyphenyl)-1,4-pentadien-3-one ≥95%
1-(Dimethylamino)-5-(3-methoxyphenyl)-1,4-pentadien-3-one
Hazard: Irritant
Formula:
C₁₄H₁₇NO₂ MW: 231.3 g/mol Melting Pt: 50…53 °C |
MDL Number:
MFCD08056608 CAS Number: 886361-10-0 |
Specification Test Results
Melting point | 50-53°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...