4-[2-(4-Chlorophenyl)acetyl]-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

042670-500MG 042670-1G 042670-5G
101856-910EA 747.69 USD
101856-910 101856-912 101856-914
4-[2-(4-Chlorophenyl)acetyl]-1H-pyrrole-2-carboxylic acid ≥95%
4-[2-(4-Chlorophenyl)acetyl]-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₃
Melting Pt: 228…230 °C
MDL Number: MFCD03001200
CAS Number: 400073-90-7

Order Now

Specification Test Results

Melting point 228-230°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR