4-Benzoyl-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

042669-500MG 042669-1G 042669-5G
101856-904EA 921.54 USD
101856-904 101856-906 101856-908
4-Benzoyl-1H-pyrrole-2-carboxylic acid ≥95%
4-Benzoyl-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₉NO₃
Melting Pt: 234…236 °C
MDL Number: MFCD03001199
CAS Number: 15372-84-6

Order Now

Specification Test Results

Melting point 234-236°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR