2-Morpholino-3-pyridinamine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-2-morpholin-4-ylpyridine
042667-500MG
042667-1G
042667-5G
101856-898EA
375.38
USD
101856-898
101856-900
101856-902
2-Morpholino-3-pyridinamine ≥95%
2-Morpholino-3-pyridinamine
Hazard: Irritant
Formula:
C₉H₁₃N₃O Melting Pt: 127…129 °C |
MDL Number:
MFCD03001243 CAS Number: 51627-47-5 |
Specification Test Results
Melting point | 127-129°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...