Ethyl-5-[(4-methylphenyl)sulfanyl]-1,2,3-thiadiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

042666-500MG 042666-1G
101856-894EA 1296.92 USD
101856-894 101856-896
Ethyl-5-[(4-methylphenyl)sulfanyl]-1,2,3-thiadiazole-4-carboxylate ≥95%
Ethyl-5-[(4-methylphenyl)sulfanyl]-1,2,3-thiadiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S₂
Melting Pt: 72…75 °C
MDL Number: MFCD00974068
CAS Number: 306977-04-8

Order Now

Specification Test Results

Melting point 72-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR