2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetonitrile ≥95%

Supplier: Matrix Scientific

042665-500MG 042665-1G 042665-5G
101856-888EA 747.69 USD
101856-888 101856-890 101856-892
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetonitrile ≥95%
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}acetonitrile

Hazard: Irritant

Formula: C₈H₄ClF₃N₂S
MW: 252.65 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD00215247
CAS Number: 306976-81-8

Order Now

Specification Test Results

Melting point 64-66°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR