2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile ≥95%

Supplier: Matrix Scientific

042663-500MG 042663-1G 042663-5G
101856-876EA 181.54 USD
101856-876 101856-878 101856-880
2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile ≥95%
2-[Cyano(2-thienyl)methyl]-6-fluorobenzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₇FN₂S
Melting Pt: 64…66 °C
MDL Number: MFCD00231908
CAS Number: 306976-61-4

Order Now

Specification Test Results

Melting point 64-66°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR