2-(4-Chlorophenyl)-4-methyl-5-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

042659-500MG 042659-1G 042659-5G
101856-852EA 83.08 USD
101856-852 101856-854 101856-856
2-(4-Chlorophenyl)-4-methyl-5-thiazolecarboxylic acid ≥95%
2-(4-Chlorophenyl)-4-methyl-5-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₈ClNO₂S
Melting Pt: 268-269°C
MDL Number: MFCD00141960
CAS Number: 54001-17-1

Order Now

Specification Test Results

Melting point 268-269°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR