3-(1,3-Benzodioxol-5-yl)-2-(3-chloropropyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042657-500MG 042657-1G 042657-5G
101856-840EA 181.54 USD
101856-840 101856-842 101856-844
3-(1,3-Benzodioxol-5-yl)-2-(3-chloropropyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%
3-(1,3-Benzodioxol-5-yl)-2-(3-chloropropyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₁₈ClNO₅
Melting Pt: 174…176 °C
MDL Number: MFCD03001229
CAS Number: 385383-35-7

Order Now

Specification Test Results

Melting point 174-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR