2-(2-Propynyloxy)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
042654-500MG
042654-1G
042654-5G
101856-824EA
140
USD
101856-824
101856-826
101856-828
2-(2-Propynyloxy)benzenecarboxylic acid ≥95%
2-(2-Propynyloxy)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₈O₃ MW: 176.17 g/mol Melting Pt: 86…87 °C |
MDL Number:
MFCD03001228 CAS Number: 55582-31-5 |
Specification Test Results
Melting point | 86-87°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...