Methyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]thio}benzoate

042653-500MG 042653-1G 042653-5G
101856-818EA 375.38 USD
101856-818 101856-820 101856-822
Methyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}benzenecarboxylate ≥95%
Methyl-2-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]sulfanyl}benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₉ClF₃NO₂S
Melting Pt: 101…102 °C
MDL Number: MFCD03001227
CAS Number: 385383-52-8

Order Now

Specification Test Results

Melting point 101-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR