4,5-Dihydroxytetrahydro-2H-imidazol-2-one ≥95%

Supplier: Matrix Scientific

042651-500MG 042651-1G 042651-5G
101856-808EA 556.92 USD
101856-808 101856-810 101856-812
4,5-Dihydroxytetrahydro-2H-imidazol-2-one ≥95%
4,5-Dihydroxytetrahydro-2H-imidazol-2-one

Hazard: Irritant

Formula: C₃H₆N₂O₃
Melting Pt: 157…160 °C
MDL Number: MFCD00186386
CAS Number: 3720-97-6

Order Now

Specification Test Results

Melting point 157-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR