Methyl-4-aminosalicylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-amino-2-hydroxybenzoate
042649-25G
042649-100G
042649-500G
101856-798EA
49.23
USD
101856-798
101856-800
101856-802
Methyl-4-aminosalicylate ≥95%
Methyl 4-aminosalicylate
Hazard: Irritant
Formula:
C₈H₉NO₃ MW: 167.18 g/mol Melting Pt: 120-123°C |
MDL Number:
MFCD00088091 CAS Number: 4136-97-4 |
Specification Test Results
Melting point | 120-123°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...