1-Methyl-1H-indazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-1-methyl-1H-indazole, 1-methyl-1H-indazole-5-amine, 1-METHYL-1H-INDAZOL-5-AMINE

042647-500MG 042647-1G
101856-788EA 1109.23 USD
101856-788 101856-790
1-Methyl-1H-indazol-5-amine ≥95%
1-Methyl-1H-indazol-5-amine

Hazard: Irritant

Formula: C₈H₉N₃
MW: 147.181 g/mol
Melting Pt: 142…144 °C
MDL Number: MFCD03305455
CAS Number: 50593-24-3

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR