2-Methyl-5-nitro-2H-indazole ≥95%

Supplier: Matrix Scientific

042645-500MG 042645-1G
101856-778EA 921.54 USD
101856-778 101856-780
2-Methyl-5-nitro-2H-indazole ≥95%
2-Methyl-5-nitro-2H-indazole

Hazard: Irritant

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Melting Pt: 130-132°C
MDL Number: MFCD00051809
CAS Number: 5228-48-8

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR