2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetohydrazide ≥95%

Supplier: Matrix Scientific

042641-500MG 042641-1G 042641-5G
101856-756EA 747.69 USD
101856-756 101856-758 101856-760
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetohydrazide ≥95%
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetohydrazide

Hazard: Irritant

Formula: C₈H₈ClF₃N₄O
MW: 268.63 g/mol
Melting Pt: 161…163 °C
MDL Number: MFCD00141298
CAS Number: 446276-13-7

Order Now

Specification Test Results

Melting point 161-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR