Ethyl-5-amino-3-(2-hydroxyethoxy)-4-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

042640-500MG 042640-1G 042640-5G
101856-750EA 375.38 USD
101856-750 101856-752 101856-754
Ethyl-5-amino-3-(2-hydroxyethoxy)-4-isoxazolecarboxylate ≥95%
Ethyl-5-amino-3-(2-hydroxyethoxy)-4-isoxazolecarboxylate

Hazard: Irritant

Formula: C₈H₁₂N₂O₅
Melting Pt: 128…130 °C
MDL Number: MFCD00172362
CAS Number: 446276-12-6

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR