Methyl-3-[5-cyano-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

042639-500MG 042639-1G 042639-5G
101856-744EA 747.69 USD
101856-744 101856-746 101856-748
Methyl-3-[5-cyano-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]-2-thiophenecarboxylate ≥95%
Methyl-3-[5-cyano-2,4-dioxo-3,4-dihydro-1(2H)-pyrimidinyl]-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₁H₇N₃O₄S
Melting Pt: 260…262 °C
MDL Number: MFCD00172924
CAS Number: 446276-11-5

Order Now

Specification Test Results

Melting point 260-262°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR