Ethyl-4-(4-methoxyphenyl)-2-oxo-6-phenyl-3-cyclohexene-1-carboxylate ≥95%

Supplier: Matrix Scientific

042632-500MG 042632-1G 042632-5G
101856-702EA 181.54 USD
101856-702 101856-704 101856-706
Ethyl-4-(4-methoxyphenyl)-2-oxo-6-phenyl-3-cyclohexene-1-carboxylate ≥95%
Ethyl-4-(4-methoxyphenyl)-2-oxo-6-phenyl-3-cyclohexene-1-carboxylate

Hazard: Irritant

Formula: C₂₂H₂₂O₄
Melting Pt: 107…108 °C
MDL Number: MFCD02031258
CAS Number: 25960-25-2

Order Now

Specification Test Results

Melting point 107-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR